Assay Details
Target Gene Details
Entrez Gene ID: | 107146 |
Gene Name: | glycine-N-acyltransferase |
Gene Aliases: |
A330009E03Rik, ACGNAT, AI195249, AI315345, CAT, GAT |
Location: |
Chr.19:12633308-12651737 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Glyat | NM_145935.3 | NP_666047.1 | ||
| AK039262.1 | BAC30297.1 | |||
| BC010799.1 | AAH10799.1 | |||
| BC024434.1 | AAH24434.1 |
More Information
Set Membership: |
|

Genomic Map