Assay Details
Target Gene Details
Entrez Gene ID: | 78825 |
Gene Name: | desumoylating isopeptidase 2 |
Gene Aliases: |
5830417C01Rik, DeSI-2, Fam152a, Pppde1 |
Location: |
Chr.1:178187231-178252598 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Desi2 | NM_024282.3 | NP_077244.1 | ||
| AK020014.1 | BAB31967.1 | |||
| AK029574.1 | BAC26520.1 | |||
| AK049579.1 | BAC33822.1 | |||
| AK146492.1 | BAE27211.1 | |||
| AK160733.1 | BAE35977.1 | |||
| AK216786.1 | ||||
| BC002200.1 | AAH02200.1 | |||
| BC046816.1 | AAH46816.1 |
More Information
Set Membership: |
|

Genomic Map