Assay Details
Target Gene Details
Entrez Gene ID: | 108154 |
Gene Name: | a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 6 |
Gene Aliases: |
5031426K13, A930019D11Rik, ADAM-TS6, b2b1879.1Clo, b2b2029Clo, b2b2182Clo, b2b2187.1Clo, b2b2228Clo |
Location: |
Chr.13:104287347-104496694 on Build GRCm38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adamts6 | NM_001081020.1 | NP_001074489.1 | ||
| AK030315.1 | ||||
| AK044528.1 |
More Information
Set Membership: |
|

Genomic Map