Assay Details
Target Gene Details
Entrez Gene ID: | 433180 |
Gene Name: | serine peptidase inhibitor, Kazal type 6 |
Gene Aliases: |
EG433180 |
Location: |
Chr.18:44071393-44083610 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spink6 | NM_001013797.1 | NP_001013819.1 | ||
| AK028032.1 | BAC25711.1 | |||
| BC147471.1 | ||||
| BC171990.1 |
More Information
Set Membership: |
|

Genomic Map