Assay Details
Target Gene Details
Entrez Gene ID: | 107971 |
Gene Name: | fibroblast growth factor receptor substrate 3 |
Gene Aliases: |
4930417B13Rik, AI449674, Frs2beta, Snt2 |
Location: |
Chr.17:47687296-47704286 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Frs3 |
Target Gene Details
Entrez Gene ID: | 381104 |
Gene Name: | prickle homolog 4 (Drosophila) |
Gene Aliases: |
Gm939 |
Location: |
Chr.17:47688474-47694958 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prickle4 | NM_001290337.1 | NP_001277266.1 |
More Information
Set Membership: |
|

Genomic Map