Assay Details
Target Gene Details
Entrez Gene ID: | 320135 |
Gene Name: | cDNA sequence BC049715 |
Gene Aliases: |
- |
Location: |
Chr.6:136828160-136841920 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BC049715 | NM_178776.3 | NP_848891.2 | ||
| AK044622.1 | 1 | 677 | BAC32007.1 | |
| AK147870.1 | 1 | 3402 | ||
| BC049715.1 | AAH49715.1 |
Target Gene Details
Entrez Gene ID: | 654818 |
Gene Name: | single-pass membrane protein with coiled-coil domains 3 |
Gene Aliases: |
C030030A07Rik |
Location: |
Chr.6:136829931-136835450 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smco3 | NM_001039558.2 | 2 | 807 | NP_001034647.2 |
| AK045581.1 | 2 | 784 | BAC32422.1 | |
| AK047766.1 | 2 | 976 | BAC33151.1 | |
| AK078719.1 | BAC37368.1 |
More Information
Set Membership: |
|

Genomic Map