Assay Details
Target Gene Details
Entrez Gene ID: | 12012 |
Gene Name: | bile acid-Coenzyme A: amino acid N-acyltransferase |
Gene Aliases: |
AI118337, AI158864, BAT |
Location: |
Chr.4:49489416-49507912 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Baat | NM_007519.3 | NP_031545.2 | ||
| AK017923.1 | BAC25534.1 | |||
| CB947692.1 | ||||
| U95215.1 | AAB58325.1 |
More Information
Set Membership: |
|

Genomic Map