Assay Details
Target Gene Details
Entrez Gene ID: | 77220 |
Gene Name: | transmembrane protein 200A |
Gene Aliases: |
C030003D03Rik |
Location: |
Chr.10:25991182-26079408 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem200a | NM_029881.3 | NP_084157.2 | ||
| AK032569.1 | BAC27929.1 | |||
| AK048673.1 | BAC33418.1 | |||
| AK165025.1 | ||||
| AK173299.1 | BAD32577.1 | |||
| BC141266.1 | ||||
| BC141267.1 |
More Information
Set Membership: |
|

Genomic Map