Assay Details
Target Gene Details
Entrez Gene ID: | 381286 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade B, member 3C |
Gene Aliases: |
1110001H02Rik, 1110013A16Rik, Scca2, Serpinb4 |
Location: |
Chr.1:107271201-107278371 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpinb3c | NM_201363.2 | NP_958751.2 | ||
| AK003650.1 | BAB22916.1 | |||
| AY367775.1 | AAR89289.1 | |||
| BC132091.1 |
More Information
Set Membership: |
|

Genomic Map