Assay Details
Target Gene Details
Entrez Gene ID: | 60531 |
Gene Name: | neuropeptide VF precursor |
Gene Aliases: |
Rfrp |
Location: |
Chr.6:50650871-50654393 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Npvf | NM_021892.1 | NP_068692.1 | ||
| AB040289.1 | BAB17673.1 | |||
| AF330058.2 | AAK94202.2 | |||
| AK138200.1 | BAE23577.1 | |||
| BC116905.1 | ||||
| BC116907.1 |
More Information
Set Membership: |
|

Genomic Map