Assay Details
Target Gene Details
Entrez Gene ID: | 93838 |
Gene Name: | DEAQ RNA-dependent ATPase |
Gene Aliases: |
2310066E11Rik |
Location: |
Chr.6:83057731-83067219 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dqx1 | NM_033606.3 | NP_291084.1 | ||
| AF318278.1 | AAK62817.1 | |||
| AK010062.1 | ||||
| BC075680.1 | AAH75680.1 |
More Information
Set Membership: |
|

Genomic Map