Assay Details
Target Gene Details
Entrez Gene ID: | 11853 |
Gene Name: | ras homolog gene family, member C |
Gene Aliases: |
AI324259, Arh9, Arhc |
Location: |
Chr.3:104789034-104794459 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rhoc | NM_001291859.1 | NP_001278788.1 | ||
| NM_007484.2 | NP_031510.2 | |||
| AK011599.1 | BAB27724.1 | |||
| AK152802.1 | BAE31507.1 | |||
| AK160650.1 | BAE35941.1 | |||
| BC004627.1 | AAH04627.1 | |||
| BY147599.1 | ||||
| BY315470.1 | ||||
| X80638.1 | CAA56682.1 |
More Information
Set Membership: |
|

Genomic Map