Assay Details
Target Gene Details
Entrez Gene ID: | 72368 |
Gene Name: | RIKEN cDNA 2310045N01 gene |
Gene Aliases: |
Mef2bnb |
Location: |
Chr.8:70139713-70147261 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2310045N01Rik | NM_001145552.1 | NP_001139024.1 | ||
| AK009829.1 | BAB26528.1 |
More Information
Set Membership: |
|

Genomic Map