Assay Details
Target Gene Details
Entrez Gene ID: | 53621 |
Gene Name: | CCR4-NOT transcription complex, subunit 4 |
Gene Aliases: |
Not4, Not4h, Not4hp |
Location: |
Chr.6:35022065-35133737 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cnot4 | NM_001164411.1 | NP_001157883.1 | ||
| NM_001164412.1 | NP_001157884.1 | |||
| NM_001164413.1 | NP_001157885.1 | |||
| NM_001164414.1 | NP_001157886.1 | |||
| NM_016877.4 | NP_058573.3 | |||
| AK009234.1 | BAB26155.1 | |||
| AK028190.1 | BAC25801.1 | |||
| AK029753.1 | ||||
| AK032248.1 | BAC27779.1 | |||
| AK075887.1 | ||||
| AK144760.1 | ||||
| AK165997.1 | BAE38508.1 | |||
| BC048243.1 | ||||
| BC058778.1 | AAH58778.1 | |||
| U71269.1 | AAD00181.1 |
More Information
Set Membership: |
|

Genomic Map