Assay Details
Target Gene Details
Entrez Gene ID: | 241134 |
Gene Name: | neuronal tyrosine-phophorylated phosphoinositide 3-kinase adaptor 2 |
Gene Aliases: |
9430031J16Rik, Jr6, Kiaa1486 |
Location: |
Chr.1:81076553-81365845 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nyap2 | NM_172849.3 | NP_766437.2 | ||
| AB429290.1 | ||||
| AK034747.1 | BAC28819.1 | |||
| AK043512.1 | ||||
| AK049778.1 | BAC33918.1 | |||
| AK137094.1 | BAE23236.1 | |||
| BC082310.1 | AAH82310.1 |
More Information
Set Membership: |
|

Genomic Map