Assay Details
Target Gene Details
Entrez Gene ID: | 100465 |
Gene Name: | MOB kinase activator 3C |
Gene Aliases: |
AW822253, D130076I06Rik, Mobkl2c |
Location: |
Chr.4:115818756-115836185 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mob3c | NM_175308.4 | NP_780517.1 | ||
| AK084014.1 | BAC39097.1 | |||
| AK148476.1 | BAE28575.1 | |||
| BC099740.1 | AAH99740.1 | |||
| BC137587.1 | ||||
| BC137589.1 |
More Information
Set Membership: |
|

Genomic Map