Assay Details
Target Gene Details
Entrez Gene ID: | 18974 |
Gene Name: | polymerase (DNA directed), epsilon 2 (p59 subunit) |
Gene Aliases: |
- |
Location: |
Chr.12:69201773-69228242 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pole2 | NM_011133.2 | NP_035263.1 | ||
| AF036898.1 | AAC40045.1 | |||
| AK011194.1 | ||||
| AK077592.1 | BAC36883.1 | |||
| AK172024.1 | BAE42782.1 | |||
| BC063772.1 | AAH63772.1 |
More Information
Set Membership: |
|

Genomic Map