Assay Details
Target Gene Details
Entrez Gene ID: | 230161 |
Gene Name: | acyl-coenzyme A amino acid N-acyltransferase 1 |
Gene Aliases: |
AI132189 |
Location: |
Chr.4:49443532-49481312 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acnat1 | NM_001164565.1 | NP_001158037.1 | ||
| DQ469311.1 |
More Information
Set Membership: |
|

Genomic Map