Assay Details
Target Gene Details
Entrez Gene ID: | 66653 |
Gene Name: | BRF2, subunit of RNA polymerase III transcription initiation factor, BRF1-like |
Gene Aliases: |
2700059M06Rik, 5730512K07Rik, AU018504, AU022602, BRFU, TFIIIB50 |
Location: |
Chr.8:27123355-27128699 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Brf2 | NM_025686.2 | NP_079962.1 | ||
| AK012459.1 | ||||
| AK012680.1 | BAB28406.1 | |||
| AK017767.1 | ||||
| AK028228.1 | BAC25827.1 | |||
| AK083122.1 | ||||
| AK151198.1 | BAE30195.1 | |||
| AK152842.1 | ||||
| BC098232.1 | AAH98232.1 |
More Information
Set Membership: |
|

Genomic Map