Assay Details
Target Gene Details
Entrez Gene ID: | 216877 |
Gene Name: | DEAH (Asp-Glu-Ala-His) box polypeptide 33 |
Gene Aliases: |
3110057P17Rik, 9430096J02Rik, Ddx33 |
Location: |
Chr.11:70984091-71004553 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dhx33 | NM_178367.4 | NP_848144.3 | ||
| AK035177.1 | BAC28969.1 | |||
| AK087592.1 | BAC39938.1 | |||
| BC052172.1 | AAH52172.1 |
More Information
Set Membership: |
|

Genomic Map