Assay Details
Target Gene Details
Entrez Gene ID: | 71388 |
Gene Name: | RIKEN cDNA 5530401A14 gene |
Gene Aliases: |
- |
Location: |
Chr.11:81860677-81894582 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 5530401A14Rik | NR_038010.1 | |||
| AK017430.1 | BAB30740.1 |
Target Gene Details
Entrez Gene ID: | 11418 |
Gene Name: | acid-sensing (proton-gated) ion channel 2 |
Gene Aliases: |
ACIC2, Accn1, BNC1, BNaC1, BNaC1a, Mdeg |
Location: |
Chr.11:80880163-81968396 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asic2 | NM_001034013.2 | NP_001029185.1 | ||
| AF348465.1 | AAK40101.1 | |||
| AK134778.1 | BAE22279.1 |
More Information
Set Membership: |
|

Genomic Map