Assay Details
Target Gene Details
Entrez Gene ID: | 24010 |
Gene Name: | IK cytokine |
Gene Aliases: |
MuRED |
Location: |
Chr.18:36744656-36757639 on Build GRCm38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ik | NM_011879.2 | NP_036009.1 | ||
| AJ006130.1 | CAA06880.1 | |||
| AK029644.1 | BAC26544.1 | |||
| AK050625.1 | BAC34349.1 | |||
| AK082356.1 | ||||
| AK087976.1 | BAC40070.1 | |||
| AK088908.1 | ||||
| AK163766.1 | BAE37486.1 | |||
| AK167238.1 | BAE39360.1 | |||
| BC014739.1 | AAH14739.1 | |||
| BC026569.1 | AAH26569.1 | |||
| BC029671.1 | AAH29671.1 | |||
| BC029694.1 | AAH29694.1 | |||
| BC033356.1 | AAH33356.1 |
More Information
Set Membership: |
|

Genomic Map