Assay Details
Target Gene Details
Entrez Gene ID: | 212307 |
Gene Name: | microtubule-associated protein, RP/EB family, member 2 |
Gene Aliases: |
AI314113, C820009F03Rik, D18Abb1e, EB1, EB2, RP1 |
Location: |
Chr.18:23752333-23893861 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mapre2 | NM_001162941.1 | NP_001156413.1 | ||
| NM_001162942.1 | NP_001156414.1 | |||
| NM_153058.4 | NP_694698.3 | |||
| AI325879.1 | ||||
| AK028823.1 | BAC26138.1 | |||
| AK038562.1 | BAC30045.1 | |||
| AK040767.1 | BAC30700.1 | |||
| AK045493.1 | BAC32393.1 | |||
| AK045904.1 | BAE43326.1 | |||
| AK046095.1 | ||||
| AK046221.1 | BAC32643.1 | |||
| AK148954.1 | BAE28700.1 | |||
| AK168834.1 | BAE40658.1 | |||
| BC025804.1 | AAH25804.1 | |||
| BC027056.1 | AAH27056.1 | |||
| BC028987.1 | AAH28987.1 | |||
| BC035254.1 | AAH35254.1 |
More Information
Set Membership: |
|

Genomic Map