Assay Details
Target Gene Details
Entrez Gene ID: | 23853 |
Gene Name: | differentially expressed in FDCP 6 |
Gene Aliases: |
2410003F05Rik, 6430538D02Rik, AV094905, Ibp, Slat, Slat2, Slat6 |
Location: |
Chr.17:28207778-28228608 on Build GRCm38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Def6 | NM_027185.3 | NP_081461.2 | ||
| AF329497.1 | AAO67354.1 | |||
| AK010356.1 | BAB26876.1 | |||
| AK038050.1 | BAE20528.1 | |||
| AK088460.1 | BAC40366.1 | |||
| AY241695.1 | AAO91768.1 | |||
| AY278770.1 | ||||
| BC120500.1 | ||||
| BC131953.1 |
More Information
Set Membership: |
|

Genomic Map