Assay Details
Target Gene Details
Entrez Gene ID: | 71952 |
Gene Name: | RIKEN cDNA 2410016O06 gene |
Gene Aliases: |
MAPJD, NO66 |
Location: |
Chr.12:83950608-83952953 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2410016O06Rik | NM_023633.3 | 1 | 393 | NP_076122.2 |
| AB041553.1 | 1 | 332 | BAA95038.1 | |
| AK080870.1 | 1 | 335 | ||
| BC138094.1 | 1 | 393 |

Genomic Map