Assay Details
Target Gene Details
Entrez Gene ID: | 17189 |
Gene Name: | myoglobin |
Gene Aliases: |
AI325109 |
Location: |
Chr.15:77015487-77050668 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mb | NM_001164047.1 | NP_001157519.1 | ||
| NM_001164048.1 | NP_001157520.1 | |||
| NM_013593.3 | NP_038621.2 | |||
| AA271284.1 | ||||
| AK137456.1 | BAE23359.1 | |||
| BC025172.1 | AAH25172.1 |
More Information
Set Membership: |
|

Genomic Map