Assay Details
Target Gene Details
Entrez Gene ID: | 245532 |
Gene Name: | acyl-CoA wax alcohol acyltransferase 2 |
Gene Aliases: |
9430062J17Rik, ARAT, Dgat2l4, WS |
Location: |
Chr.X:100399336-100494221 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Awat2 | NM_001290395.1 | NP_001277324.1 | ||
| AK034920.1 | BAC28882.1 |
Target Gene Details
Entrez Gene ID: | 408193 |
Gene Name: | OTU domain containing 6A |
Gene Aliases: |
EG408193, Hshin6 |
Location: |
Chr.X:100429013-100429885 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Otud6a | NM_001163191.1 | 1 | 98 | NP_001156663.1 |
| BC153817.1 | 1 | 98 |
More Information
Set Membership: |
|

Genomic Map