Assay Details
Target Gene Details
Entrez Gene ID: | 103080 |
Gene Name: | septin 10 |
Gene Aliases: |
4921515A04Rik, 9430099J10Rik, AA408298, AI874685 |
Location: |
Chr.10:59141627-59221847 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sept10 | NM_001024910.3 | NP_001020081.2 | ||
| NM_001024911.2 | NP_001020082.1 | |||
| AK076569.1 | BAC36397.1 | |||
| BC053752.1 | AAH53752.1 |
More Information
Set Membership: |
|

Genomic Map