Assay Details
Target Gene Details
Entrez Gene ID: | 18741 |
Gene Name: | paired-like homeodomain transcription factor 2 |
Gene Aliases: |
9430085M16Rik, Brx1, Brx1b, Munc30, Otlx2, Ptx2, Rieg |
Location: |
Chr.3:129199878-129219594 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pitx2 | NM_001042502.2 | NP_001035967.1 | ||
| NM_001042504.2 | NP_001035969.1 | |||
| NM_001286942.1 | NP_001273871.1 | |||
| NM_001287048.1 | NP_001273977.1 | |||
| NM_011098.4 | NP_035228.2 | |||
| AB006320.1 | BAA75247.1 | |||
| AB006321.1 | BAA75248.1 | |||
| AF048723.1 | AAC40086.1 | |||
| AF048724.1 | AAC40087.1 | |||
| AF201091.1 | AAF44618.1 | |||
| AK034846.1 | 1 | 1255 | ||
| AK035083.1 | BAC28939.1 | |||
| AM940438.1 | ||||
| AM940439.1 | ||||
| BC075660.1 | AAH75660.1 | |||
| U80010.1 | AAC53119.1 | |||
| U80011.1 | AAC53120.1 | |||
| U80036.1 | AAB38505.1 |
More Information
Set Membership: |
|

Genomic Map