Assay Details
Target Gene Details
Entrez Gene ID: | 27425 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial F0 complex, subunit G |
Gene Aliases: |
4933437C06Rik |
Location: |
Chr.9:44913248-44920742 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
More Information
Set Membership: |
|

Genomic Map