Assay Details
Target Gene Details
Entrez Gene ID: | 14051 |
Gene Name: | eyes absent 4 homolog (Drosophila) |
Gene Aliases: |
B130023L16Rik |
Location: |
Chr.10:23102962-23350856 on Build GRCm38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eya4 | NM_010167.4 | NP_034297.2 | ||
| AK041625.1 | BAC31011.1 | |||
| BC120898.1 | ||||
| BC120899.1 | ||||
| Y17115.1 | CAA76637.1 |
More Information
Set Membership: |
|

Genomic Map