Assay Details
Target Gene Details
Entrez Gene ID: | 70638 |
Gene Name: | family with sequence similarity 189, member A1 |
Gene Aliases: |
5730507A09Rik, AW047451 |
Location: |
Chr.7:64756091-65156564 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam189a1 | NM_183087.4 | NP_898910.2 | ||
| AK172974.1 | BAD32252.1 | |||
| CJ060159.1 |
Target Gene Details
Entrez Gene ID: | 66647 |
Gene Name: | necdin-like 2 |
Gene Aliases: |
5730494G16Rik, AI642138, BB044375, HCA4, Mageg1, mage-g1 |
Location: |
Chr.7:64871650-64873040 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndnl2 | NM_023239.4 | 1 | 165 | NP_075728.1 |
| AF319979.1 | AAK01207.1 | |||
| AK010294.1 | 1 | 126 | BAB26830.1 | |
| AK017727.1 | 1 | 167 | BAB30899.1 | |
| AK018250.1 | 1 | 126 | BAB31133.1 | |
| AK049759.1 | 1 | 145 | BAC33907.1 | |
| AK076471.1 | 1 | 151 | BAC36358.1 | |
| BC034892.1 | 1 | 90 | AAH34892.1 | |
| BC092289.1 | 1 | 127 |
More Information
Set Membership: |
|

Genomic Map