Assay Details
Target Gene Details
Entrez Gene ID: | 20760 |
Gene Name: | small proline-rich protein 2F |
Gene Aliases: |
- |
Location: |
Chr.3:92365187-92366442 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sprr2f | NM_011472.2 | NP_035602.1 | ||
| AY158990.1 | AAN86827.1 |
More Information
Set Membership: |
|

Genomic Map