Assay Details
Target Gene Details
Entrez Gene ID: | 23893 |
Gene Name: | gremlin 2 homolog, cysteine knot superfamily (Xenopus laevis) |
Gene Aliases: |
Gremlin2, Prdc |
Location: |
Chr.1:174833785-174921915 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grem2 | NM_011825.1 | 2 | 983 | NP_035955.1 |
| AB011030.1 | 2 | 983 | BAA29038.1 | |
| AK142008.1 | 2 | 985 | BAE24912.1 | |
| AK161827.1 | 2 | 963 | BAE36594.1 | |
| BC079905.1 | 2 | 964 | AAH79905.1 |

Genomic Map