Assay Details
Target Gene Details
Entrez Gene ID: | 68018 |
Gene Name: | collagen, type IV, alpha 3 (Goodpasture antigen) binding protein |
Gene Aliases: |
2810404O15Rik, 9230101K08Rik, AU016711, CERT, GPBP |
Location: |
Chr.13:96542735-96640167 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Col4a3bp | NM_001164222.1 | NP_001157694.1 | ||
| NM_023420.2 | NP_075909.1 | |||
| AF232932.1 | AAG42048.1 | |||
| AF232934.1 | AAG42050.1 | |||
| AK012989.1 | BAB28581.1 | |||
| AK030305.1 | BAC26888.1 | |||
| AK038061.1 | BAC29927.1 | |||
| AK038803.1 | ||||
| AK047100.1 | BAC32962.1 | |||
| AK080958.1 | BAC38095.2 | |||
| AK163624.1 | BAE37427.1 | |||
| BC016197.1 | AAH16197.1 |
More Information
Set Membership: |
|

Genomic Map