Assay Details
Target Gene Details
Entrez Gene ID: | 76080 |
Gene Name: | tocopherol (alpha) transfer protein-like |
Gene Aliases: |
3110080A02Rik, 5830472M02Rik |
Location: |
Chr.2:163602281-163619013 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttpal | NM_029512.2 | NP_083788.2 | ||
| NM_181734.3 | NP_859423.2 | |||
| AK018052.1 | ||||
| AK033600.1 | BAC28381.1 | |||
| AK039450.1 | BAC30352.1 | |||
| AK043791.1 | BAC31654.1 | |||
| AK052794.1 | ||||
| AK081680.1 | BAC38289.1 | |||
| AK156641.1 | BAE33788.1 | |||
| BC052389.1 | ||||
| BY717985.1 | ||||
| CJ183358.1 |
More Information
Set Membership: |
|

Genomic Map