Assay Details
Target Gene Details
Entrez Gene ID: | 22238 |
Gene Name: | UDP glucuronosyltransferase 2 family, polypeptide B5 |
Gene Aliases: |
AI118071, M-1, Udpgt-3, Udpgt2b5, Ugt2b17 |
Location: |
Chr.5:87124946-87140340 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ugt2b5 | NM_009467.3 | NP_033493.3 | ||
| AK136480.1 | BAE22997.1 | |||
| BC028262.1 | AAH28262.1 | |||
| X06358.1 | CAA29657.1 |
More Information
Set Membership: |
|

Genomic Map