Assay Details
Target Gene Details
Entrez Gene ID: | 102902673 |
Gene Name: | predicted gene 21992 |
Gene Aliases: |
Rbm14-rbm4 |
Location: |
Chr.19:4784293-4811634 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm21992 | NM_001290127.1 | NP_001277056.1 | ||
| NM_001290128.1 | NP_001277057.1 | |||
| AK087488.1 | BAC39895.1 | |||
| EU287940.1 | ||||
| EU287941.1 |
Target Gene Details
Entrez Gene ID: | 56275 |
Gene Name: | RNA binding motif protein 14 |
Gene Aliases: |
1300007E16Rik, PSP2, Sytip, p16, p16K |
Location: |
Chr.19:4800566-4811634 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbm14 | NM_019869.3 | NP_063922.2 | ||
| AK004928.1 | BAB23678.1 | |||
| AK050945.1 | ||||
| AK088201.1 | BAC40206.1 | |||
| AK153230.1 | BAE31823.1 | |||
| AK167503.1 | BAE39579.1 | |||
| BC010294.1 | AAH10294.1 |
More Information
Set Membership: |
|

Genomic Map