Assay Details
Target Gene Details
Entrez Gene ID: | 13142 |
Gene Name: | D-amino acid oxidase |
Gene Aliases: |
AI987963, DAAO, DAMOX, Dao-1, Dao1 |
Location: |
Chr.5:113999636-114025675 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dao | NM_001286397.1 | NP_001273326.1 | ||
| NM_010018.3 | NP_034148.2 | |||
| AK134813.1 | BAE22296.1 | |||
| BB843516.1 | ||||
| BC015269.1 | AAH15269.1 | |||
| BE651085.1 | ||||
| M32299.1 | AAA39367.1 |
More Information
Set Membership: |
|

Genomic Map