Assay Details
| Gene Symbol: Olfr1180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146918.2 | NP_667129.2 | 1 | 483 | 70 | |
| GenBank mRNA | BC120811.1 | - | 1 | 483 | 40129734 | 70 |
| BC120813.1 | - | 1 | 483 | 40129736 | 70 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2098930 | BC120811.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2098930 | BC120813.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2098930 | NM_146918.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145392 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2098930 | BC120811.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2098930 | BC120813.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2098930 | NM_146918.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P466258 | BC120811.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P466258 | BC120813.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P466258 | NM_146918.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145392 | - | - |
| Whole Mouse Genome G4122A | A_51_P466258 | BC120811.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P466258 | BC120813.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P466258 | NM_146918.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140256 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21409649 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02235290 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409687 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409693 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02188841 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409678 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409686 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409711 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01555375 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409660 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409651 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409689 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409653 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409665 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409701 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409684 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409707 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409695 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409662 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409673 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409647 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409640 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409642 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409664 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409654 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409669 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409716 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409649 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409681 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01217058 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409690 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409704 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409656 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409679 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409671 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21409709 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10484742 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10484743 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10484744 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10484741 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 814063 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6550551 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006550551 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2649883 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2649883 | - | - |
| mouse-6 v1.1 (Illumina) | scl4299.1.1_130-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6550551 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl4299.1.1_130-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6550551 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2649883 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2649883 | - | - |

Olfr1180
Genomic Map