Assay Details
| Gene Symbol: LOC4342780 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792281.2 | XP_015647767.1 | 3 - 4 | 719 | 155 | |
| XM_015792282.2 | XP_015647768.1 | 2 - 3 | 787 | 155 | ||
| GenBank mRNA | AK103406.1 | - | 3 - 4 | 674 | 155 | |
More Information
Set Membership: |
|

LOC4342780
Genomic Map