Assay Details
| Gene Symbol: LOC4324704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792773.2 | XP_015648259.1 | 1 | 1094 | 76 | |
| GenBank mRNA | AK061237.1 | - | 1 | 1047 | 76 | |
More Information
Set Membership: |
|

LOC4324704
Genomic Map