Assay Details
| Gene Symbol: Hmbs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013168.2 | NP_037300.2 | 6 - 7 | 475 | 99 | |
| XM_006242896.1 | XP_006242958.1 | 6 - 7 | 561 | 99 | ||
| XM_008766148.1 | XP_008764370.1 | 5 - 6 | 645 | 99 | ||
| XM_008766149.1 | XP_008764371.1 | 5 - 6 | 582 | 99 | ||
| GenBank mRNA | BC088162.1 | - | 6 - 7 | 475 | 7375228 | 99 |
| FQ226062.1 | - | 6 - 7 | 398 | 99 | ||
| FQ231091.1 | - | 6 - 7 | 459 | 99 | ||
| FQ234870.1 | - | 4 - 5 | 631 | 99 | ||
| X06827.1 | - | 6 - 7 | 350 | 99 | ||
| Y12006.1 | - | 6 - 7 | 345 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hmbs
Genomic Map