Assay Details
Gene Family ID: | MIPF0000071, mir-302 |
Mature miRNA Sequence: | UAAGUGCUUCCAUGUUUCAGUGG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-302c-3p | MIMAT0000717 | |
| Bovine | bta-miR-302c | MIMAT0009281 | |
| Callithrix jacchus | cja-miR-302c | MIMAT0039615 | |
| Chicken | gga-miR-302c-3p | MIMAT0003359 | |
| Dasypus novemcinctus | dno-miR-302c-3p | MIMAT0047796 | |
| Horse | eca-miR-302c | MIMAT0012913 | |
| View More Rows | |||
| Pan troglodytes | ptr-miR-302c | MIMAT0008088 | |
| Pongo pygmaeus | ppy-miR-302c | MIMAT0015818 | |
| Rabbit | ocu-miR-302c-3p | MIMAT0036319 | |
| Rhesus monkey | mml-miR-302c | MIMAT0006261 | |
Stem-loop Details
Human
Bovine
Callithrix jacchus
Chicken
Dasypus novemcinctus
Horse
Pan troglodytes
Pongo pygmaeus
Rabbit
Rhesus monkey
More Information
Related Products
miRBase Alias:
Human: hsa-miR-302c (v17)
Gallus gallus: gga-miR-302c (v18)
Gallus gallus: gga-miR-302c (v18)

Genomic Map