Assay Details
Amplicon Length: | 85 |
Chromosome Location: | Chr. 4 - 112648706 - 112648706 [-] on 112648706 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-302a-5p | MIMAT0000683 |
| Human | hsa-miR-302a-3p | MIMAT0000684 |
| Human | hsa-miR-302b-5p | MIMAT0000714 |
| Human | hsa-miR-302b-3p | MIMAT0000715 |
| Human | hsa-miR-302c-5p | MIMAT0000716 |
| Human | hsa-miR-302c-3p | MIMAT0000717 |
Stem-loop Details
Human
More Information
Related Products
miRBase Alias:
Human: hsa-mir-302a (v21)
Human: hsa-mir-302c (v21)
Human: hsa-mir-302c (v21)

Genomic Map