Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | AUCUGGAGGUAAGAAGCACUUU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-516b-5p | MIMAT0002859 |
| Pan troglodytes | ptr-miR-516b | MIMAT0008183 |
| Pongo pygmaeus | ppy-miR-516b | MIMAT0015947 |
| Rhesus monkey | mml-miR-516b | MIMAT0012778 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
More Information
Related Products
miRBase Alias:
Human: hsa-miR-516b (v17)
Macaca mulatta: mml-miR-516 (v18)
Macaca mulatta: mml-miR-516 (v18)

Genomic Map