Assay Details
Amplicon Length: | 113 |
Chromosome Location: | Chr. 19 - 53737247 - 53737247 [+] on 53737247 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-516b-5p | MIMAT0002859 |
| Human | hsa-miR-516b-3p | MIMAT0002860 |
Stem-loop Details
Human

Genomic Map