Assay Details
Gene Family ID: | MIPF0000496, mir-589 |
Mature miRNA Sequence: | UCAGAACAAAUGCCGGUUCCCAGA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-589-3p | MIMAT0003256 |
Stem-loop Details
Human
More Information
Related Products
miRBase Alias:
Human: hsa-miR-589* (v17)
Human: hsa-miR-589 (v9.2)
Human: hsa-miR-589 (v9.2)

Genomic Map