Assay Details
Amplicon Length: | 67 |
Chromosome Location: | Chr. 7 - 5496118 - 5496118 [-] on 5496118 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-589-5p | MIMAT0004799 |
| Human | hsa-miR-589-3p | MIMAT0003256 |
Stem-loop Details
Human

Genomic Map