Assay Details
Gene Family ID: | MIPF0000167, mir-370 |
Mature miRNA Sequence: | GCCUGCUGGGGUGGAACCUGGU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number | |
---|---|---|---|
Human | hsa-miR-370-3p | MIMAT0000722 | |
Mouse | mmu-miR-370-3p | MIMAT0001095 | |
Rat | rno-miR-370-3p | MIMAT0003122 | |
Bovine | bta-miR-370 | MIMAT0009300 | |
Dog | cfa-miR-370 | MIMAT0009889 | |
Eptesicus fuscus | efu-miR-370 | MIMAT0034931 | |
View More Rows | |||
Guinea pig | cpo-miR-370-3p | MIMAT0047202 | |
Horse | eca-miR-370 | MIMAT0013142 | |
Pan paniscus | ppa-miR-370 | MIMAT0049048 | |
Pan troglodytes | ptr-miR-370 | MIMAT0008120 | |
Pig | ssc-miR-370 | MIMAT0025373 | |
Pongo pygmaeus | ppy-miR-370 | MIMAT0015857 | |
Rabbit | ocu-miR-370-3p | MIMAT0048405 | |
Rhesus monkey | mml-miR-370-3p | MIMAT0006295 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Dog
Eptesicus fuscus
Guinea pig
Horse
Pan paniscus
Pan troglodytes
Pig
Pongo pygmaeus
Rabbit
Rhesus monkey

More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool AMegaplex™ RT Primers, Human Pool A
Related Products
miRBase Alias:
Human: hsa-miR-370 (v19)
Mouse: mmu-miR-370 (v17)
Rat: rno-miR-370 (v18)
Macaca mulatta: mml-miR-370 (v19)
Mouse: mmu-miR-370 (v17)
Rat: rno-miR-370 (v18)
Macaca mulatta: mml-miR-370 (v19)
